Learn More
Cole-Parmer™ Spark Bunsen Burner Lighter
Use this well-constructed safety spark lighter for all your burner ignition needs. Cole-Parmer™ Spark Bunsen Burner Lighter uses a large renewable flint that produces up to 1500 ignitions.
Brand:  Cole-Parmer™ 36136-50
15.20 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Product Code. 12200379
  • 16.00€ / Each
Estimated Shipment: 04-07-2025
Description
Use this well-constructed safety spark lighter for all your burner ignition needs. Cole-Parmer™ Spark Bunsen Burner Lighter uses a large renewable flint that produces up to 1500 ignitions.
  • Special spark hood protects the flint.
  • Gas pocket forms beneath the hood, ensuring instant lighting.
  • Lighter incorporates new larger, renewable flint that produces up to 1500 ignitions.
Specifications
Specifications
Gas Spark Lighter
(5) Flints
1 Ea.
Certificates
View accessory products
Product Title
Cole-Parmer™ Spark Bunsen Burner Lighter > Sparking Lighter