Learn More
BRAND™ Special Laboratory PVC Tubing
Special Laboratory Tubing constructed of polyvinyl chloride.
207.00€ - 390.00€
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
15241389
|
BRAND™
143270 |
6 mm |
207.00€
20m |
In Stock
Log in to see stock. |
|||||
15251389
|
BRAND™
143275 |
7 mm |
302.00€
20m |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
15261389
|
BRAND™
143280 |
8 mm |
322.00€
20m |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
15271389
|
BRAND™
143285 |
10 mm |
390.00€
20m |
In Stock
Log in to see stock. |
|||||
Description
- Constructed of PVC
- Available in a variety of diameters
Specifications
Tubing | |
PVC |