Learn More
3M™ Aura™ Specialty Respirator, Series 9300
Lightweight and foldable. The 3M Aura Particulate Respirator uses a unique three-panel design for greater comfort and easy communication.
98.00€ - 2195.00€
Specifications
Certifications/Compliance (Europe) | EN149:2001 + A1:2009 |
---|---|
Design Type | Cup/Molded |
Disposable | Single-use |
Product Type | Particulate Respirator with Nuisance Organic Vapor Relief |
Size | Standard |
Product Code | Brand | Filter Efficiency | With Exhalation Valve | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Efficiency | With Exhalation Valve | Color | Price | Quantity & Availability | |||||
12352399
|
3M™
7000034739 |
FFP1 | Yes | Gray |
98.00€
Pack of 10 |
In Stock
Log in to see stock. ![]() |
|||||
11763373
|
3M™
7000034737 |
FFP1 | No | Blue | N/A | ||||||
11797564
|
3M™
7000034738 |
FFP2 | Yes | Blue |
137.00€
Pack of 10 |
In Stock
Log in to see stock. ![]() |
|||||
15263059
|
3M™
7000052909 |
FFP3 | Yes | White |
2195.00€
Pack of 180 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
12352389
|
3M™
7000034740 |
FFP1 | No | Gray |
162.00€
Pack of 20 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
|||||
Description
Fits a wide range of face shapes and sizes- Low breathing resistance filter technology for easier breathing
- Ingenious three-panel design accommodates greater facial movement during speech for more comfortable wear
- An embossed top panel to reduce fogging of eyewear
- A sculpted nose panel for improved compatibility with eyewear
- An innovative chin tab for ease of donning and adjustment of the respirator on the face
- Supplied in hygienic packaging to help protect the respirator from contamination before use, also allows practical storage and dispensing in the workplace.
- Cool Flow™ valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
Specifications
EN149:2001 + A1:2009 | |
Single-use | |
Standard | |
Yes | |
Non-sterile |
Cup/Molded | |
Particulate Respirator with Nuisance Organic Vapor Relief | |
Electret (polyester, polypropylene, and carbon) | |
Cup | |
No |
Product Title
3M™ Aura™ Specialty Respirator, Series 9300