Learn More
3M™ Specialty Particulate Respirators 9000 Cup Series
Speciality particulate respirators provide lightweight and comfortable FFP1, FFP2 and FFP3 breathing protection against organic vapours, acid gases, ozone, hydrogen fluoride, and nuisance odours.
Brand:  3M™ 7000088787
48.50 EUR valid until 2025-06-08
Use promo code "24131" to get your promotional price.
Product Code. 15759837
  • 91.00€ / Pack of 10
Gear Up for Safety
Use promo code 24131 at checkout
View Available Promotions
Estimated Shipment: 15-07-2025
Description
  • Provides FFP1 (NPF 4), FFP2 (NPF 12), and FFP3 (NPF 50) protection against dusts and mists
  • Activated carbon layer helps guard against nuisance odours and, in some models, low levels of organic vapours, acid gases and ozone (NPF 10)
  • Convex shape, nose clip and twin strapping helps provide a comfortable fit across a broad range of face shapes
  • Lightweight, cup-shaped designs are sturdy and collapse resistant
  • Braided, colour-coded headbands help you identify the protection level offered: yellow for FFP1, blue for FFP2 and red for FFP3
  • Good fit over a wide range of face sizes provided by convex shape, nose clip and twin-strap design
  • Designed to work well with 3M Eyewear and Hearing Protection products
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gray
Particulate Respirator with Nuisance Organic Vapor Relief
<Hgv for ozone , Up to 10 x Hgv for particles
No
FFP2
Yes
Non-sterile
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Days
    31
    Hours
    15
    Minutes
    38
    Seconds
    52
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
3M™ Specialty Particulate Respirators 9000 Cup Series > Model: 9922; Category: Disposable Respirators; Filter Type: