Learn More
Bollé Safety SPHERE Spare Lens, Clear Product Code.: 12669170
Estimated Shipment: 09-10-2025
Only null left
Add to basket
Bollé Safety SPHERE Spare Lens, Clear
Product Code. 12669170
Product Code. 12669170
Brand: Bollé Safety FASPHERPI
Only null left
Estimated Shipment: 09-10-2025
Add to basket
Only null left
Add to basket
Spare lens for SPERE face shield
  • Protection against electrical short circuit arcs
  • 20.5 x 39.4 cm PC shield
  • Resistant to high energy impact (A marking)
  • TRUSTED_SUSTAINABILITY
    Specifications
    Thickness (Metric) 2 mm
    Disposable Reusable
    Lens Material Polycarbonate
    Product Content Correction
    Product Title
    Bollé Safety SPHERE Spare Lens, Clear >