Learn More
R&D Systems™ Cultrex Spheroid Invasion Extracellular Matrix
R&D Systems™ Spheroid Invasion Matrix is for spheroid invasion assays and offers a standardized, three-dimensional, high content format for quantitating the degree to which invasive cells penetrate a barrier, consisting of basement membrane components.
Brand: R&D Systems™ 3500-096-03
Additional Details : Weight : 0.58200kg
Description
- Dispense into working aliquots if needed; repeated freeze-thaws will destroy product integrity.
- Source: Murine Engelbreth-Holm-Swarm (EHS) tumor and bovine extensor tendon.
Specifications
Product is stable for a minimum of 3 months from date of shipment. Store at -80° C; keep frozen. Dispense into working aliquots if needed; repeated freeze-thaws will destroy product integrity. | |
6 mL | |
Product is stable for a minimum of 3 months from date of shipment. |