Learn More
Heidolph™ Spider with Distillation Sleeves
For use with rotary evaporators
Brand:  Heidolph™ 15-300-005-11
 View more versions of this product
Product Code. 10466671
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Spider come with a standard NS 29/32 joint size but are also available with NS 24/29 ground joints.
Specifications
Specifications
Glass
Rotary evaporators
20 mL (per tube)
Spider with Distillation Sleeve
20 distillation sleeves