Learn More
Uvex Sport Occupational Black PU 65958 Low Shoe - Width 11
Offers first-class wearer comfort; low weight, climate-optimizes high-tech materials and ergonomic outsole
Brand:  Uvex 6595848
118.63 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17212831
  • 135.00€ / 1 pair
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Uvex 1 Sport Shoe 6595.8 O1 FO SRC
  • Modern, ultra-lightweight and flexible O1 occupational shoe without toe cap
  • Made from synthetic materials, suitable for people allergic to chrome
  • All sole materials are free of silicones plasticizers and other paint wetting impairment substances
  • Meets ESD specification with leakage resistance less than 35 megohms
  • Ergonomically designed outsole comprising dual-density polyurethane with excellent slip resistance
  • Made with new developed technology for exceptional wearer comfort, climate-optimizes and breathable materials
  • Virtually seam-free construction made from high-tech materials to eliminate pressure points
  • Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
  • Soft, padding on collar and tongue
  • For light applications
  • Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
  • Article No. 6595.8
  • Standard EN ISO 20347:2012 O1 FO SRC
  • Sizes: 35 to 52
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
48
Plain Toe
EN ISO 20347:2012
Standard Width
Yes
O1
Textile
Dual-density polyurethane (PU/PU)
Reusable
Unisex
Uvex 1
No
SRC