Learn More
BRAND™ Spotting Tile
12 polished cavities
Brand: BRAND™ 473500
99.55 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Spotting Tile | |
Soda Lime Glass | |
1 | |
130 x 100 x 6 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
BRAND™ Spotting Tile > Dimensions (L x W x H) Exterior: 130 x 100 x 6 mm