missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR 8278, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Rev-Erbα antagonist; inhibits Rev-Erbα transcriptional repression (EC50 = 0.47 μM). Blocks activity of Rev-Erbα agonist GSK 4112 (Cat. No. 3663) in HEK293 cells. Increases expression of glucose-regulating genes, G6Pase and PEPCK in HepG2 cells.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 361.48 |
Percent Purity | >99% |
Chemical Name or Material | SR 8278 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction