Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML E2
Adjusted to Meet or Exceed OIML E2 Tolerances
36.50€ - 2080.00€
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16652342
|
Ohaus™
30518004 |
1 mg |
36.50€
Each |
In Stock
Log in to see stock. |
|||||
16662342
|
Ohaus™
30518005 |
2 mg |
36.50€
Each |
In Stock
Log in to see stock. |
|||||
16672342
|
Ohaus™
30518006 |
5 mg |
36.50€
Each |
In Stock
Log in to see stock. |
|||||
16682342
|
Ohaus™
30518007 |
10 mg |
36.50€
Each |
In Stock
Log in to see stock. |
|||||
16692342
|
Ohaus™
30518008 |
20 mg |
36.50€
Each |
In Stock
Log in to see stock. |
|||||
16602352
|
Ohaus™
30518009 |
50 mg |
36.50€
Each |
In Stock
Log in to see stock. |
|||||
16612352
|
Ohaus™
30518010 |
100 mg |
36.50€
Each |
In Stock
Log in to see stock. |
|||||
16622352
|
Ohaus™
30518011 |
200 mg |
36.50€
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
16632352
|
Ohaus™
30518012 |
500 mg |
36.50€
Each |
In Stock
Log in to see stock. |
|||||
16642352
|
Ohaus™
30518013 |
1 g |
72.75€
Each |
In Stock
Log in to see stock. |
|||||
16652352
|
Ohaus™
30518014 |
2 g |
76.50€
Each |
In Stock
Log in to see stock. |
|||||
16662352
|
Ohaus™
30518015 |
5 g |
81.50€
Each |
In Stock
Log in to see stock. |
|||||
16672352
|
Ohaus™
30518016 |
10 g |
84.25€
Each |
In Stock
Log in to see stock. |
|||||
16682352
|
Ohaus™
30518017 |
20 g |
97.25€
Each |
In Stock
Log in to see stock. |
|||||
16692352
|
Ohaus™
30518018 |
50 g |
112.00€
Each |
In Stock
Log in to see stock. |
|||||
16602362
|
Ohaus™
30518019 |
100 g |
120.00€
Each |
In Stock
Log in to see stock. |
|||||
16612362
|
Ohaus™
30518020 |
200 g |
151.00€
Each |
In Stock
Log in to see stock. |
|||||
16622362
|
Ohaus™
30518021 |
500 g |
253.00€
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
16632362
|
Ohaus™
30518022 |
1 kg |
367.00€
Each |
In Stock
Log in to see stock. |
|||||
16642362
|
Ohaus™
30518023 |
2 kg |
585.00€
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
16652362
|
Ohaus™
30518024 |
5 kg |
855.00€
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
16662362
|
Ohaus™
30518025 |
10 kg |
1240.00€
Each |
In Stock
Log in to see stock. |
|||||
16672362
|
Ohaus™
30518026 |
20 kg |
2080.00€
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
Description
- Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
No Certificate | |
Protective Case |