Learn More
Fisherbrand™ Stainless Steel Covers for Transsonic Analogous Ultrasonic Units
A safe and durable cover to complete your ultrasonic cleaning units. Fisherbrand™ Stainless Steel Covers for Transsonic Analogous Ultrasonic Units are strong, carefree and meet high safety standards.
Brand: Fisherbrand™ 2070550000
This item is not returnable.
View return policy
Description
- Easy to clean
- Clean-cut, smooth surfaces
- Meets high user demands for hygiene
- Available in a variety of styles to fit specific models - rectangular/square, round, with beaker holes or without
Specifications
Lid | |
No beaker holes | |
Rectangular/Square |
Transonic Analogous Ultrasonic Unit, T 1040/H | |
Stainless Steel |