Learn More
Fisherbrand™ Stainless Steel Covers for Transsonic Analogous Ultrasonic Units
A safe and durable cover to complete your ultrasonic cleaning units. Fisherbrand™ Stainless Steel Covers for Transsonic Analogous Ultrasonic Units are strong, carefree and meet high safety standards.
Brand:  Fisherbrand™ 2070290000
 View more versions of this product
Product Code. 10212192
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
  • Easy to clean
  • Clean-cut, smooth surfaces
  • Meets high user demands for hygiene
  • Available in a variety of styles to fit specific models - rectangular/square, round, with beaker holes or without
Specifications
Specifications
Lid
Holes for 1 beaker
Rectangular/Square
Transonic Analogous Ultrasonic Unit, T 420, T 420/H