Learn More
Grant Instruments™ Stainless Steel Empty Tank
Empty stainless-steel tank with drain tap.
Brand: Grant Instruments™ ST12
This item is not returnable.
View return policy
Description
Empty stainless-steel tank with drain tap.
- Stainless Steel construction
- With drain tap
Specifications
12 L | |
Tank | |
Bridge Plate | |
Stainless Steel |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Grant Instruments™ Stainless Steel Empty Tank > Volume: 12L