Learn More
Stainless Steel foil, 0.025mm (0.001in) thick, Type 304, Thermo Scientific Chemicals
38.40€ - 83.60€
Specifications
Chemical Name or Material | Stainless Steel |
---|---|
Color | Silver |
Name Note | Type 304 |
Appearance | Silver -colored foil |
Molecular Formula | Fe:Cr:Ni; 70:19:11 wt% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11328698
|
Thermo Scientific Alfa Aesar
041580.FF |
25 x 25 mm |
38.40€
25mm |
In Stock
Log in to see stock. |
|||||
11460401
|
Thermo Scientific Alfa Aesar
041580.FI |
50 x 50 mm |
57.20€
Each |
In Stock
Log in to see stock. |
|||||
11338698
|
Thermo Scientific Alfa Aesar
041580.GH |
100 x 100 mm |
83.60€
Each |
In Stock
Log in to see stock. |
|||||
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel | |
Type 304 | |
Fe:Cr:Ni; 70:19:11 wt% | |
It is insoluble in water. | |
Foil |
Silver | |
Silver -colored foil | |
MFCD00148499 | |
Odorless |
Safety and Handling
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only