Learn More
Bochem™ Stainless Steel Hook Connector Bosshead
Manufactured from 18/10 stainless steel. BOCHEM™ Stainless Steel Hook Connector Bosshead includes an M6 heart screw.
Brand:  Bochem™ 5304
Product Code. 10170461
  • 30.10€ / Each
Only null left
Estimated Shipment: 20-06-2025
Description
  • 18/10 Stainless steel
  • Includes heart screw M6
Specifications
Specifications
1 Ea.
Stainless Steel
16.5 mm
Hook Connector
16.5 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Stainless Steel Hook Connector Bosshead > Dia.: 16.5mm, Heart Screw M6