Learn More
Fisherbrand™ Stainless Steel Immersion Basket with Handles
Facilitates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Stainless Steel Immersion Baskets with Handles are available for a range of cleaner capacities.
Brand:  Fisherbrand™ 1001243
Product Code. 11709118
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • Mesh bottom
  • Two handles, one on each end
  • Functional and modern design
Specifications
Specifications
Basket
Ultrasonic Bath S 150