Learn More
Smeg™ Stainless Steel Instrument Basket
Designed for use with Smeg™ glassware washers. Smeg Stainless Steel Instrument Baskets are suitable for medium-sized instruments.
246.00€
Products 1
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
13287949
View Documents
Smeg™
CSK2
225 x 450 x 50mm
246.00€
Each
Estimated Shipment: 14-02-2025
Description
  • Baskets with stainless steel handles, suitable for medium-sized instruments
  • Mesh openings of 5 x 5mm
  • Dimensions: 225 x 450 x 50mm