Learn More
Usbeck Laborgeraete™ Stainless Steel Jacks
154.30€ - 175.00€
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
10629531
|
Usbeck Laborgeraete™
5250 |
130 x 160 mm |
154.30€
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
10448191
|
Usbeck Laborgeraete™
5260 |
200 x 200 mm |
175.00€
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
Specifications
Stainless Steel | |
Jack | |
1 Ea. |