Learn More
Bochem™ Stainless Steel Lid
Bochem™ Stainless Steel Lid for use with 20, 25, and 100 Liter Stainless Steel containers.
Brand:  Bochem™ 8357
 View more versions of this product
Product Code. 10672112
  • 281.00€ / Each
Estimated Shipment: 26-06-2025
Description
  • Stainless Steel
  • For use with containers
Specifications
Specifications
250mm
Stailness Steel Container
Stainless Steel (18/10)
Product Title
Bochem™ Stainless Steel Lid > Diameter: 450mm