Learn More
Usbeck Laborgeraete™ Stainless Steel Measuring Cup
Brand:  Usbeck Laborgeraete™ 4402
Product Code. 10340802
  • 42.60€ / Each
Estimated Shipment: 09-07-2025
Description
Order Info
CapacityMetric
Specifications
Specifications
Stainless Steel
1500 mL
160 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Usbeck Laborgeraete™ Stainless Steel Measuring Cup > Capacity: 1500mL