Specifications
Pastry Bowl | |
For making mixtures, mixing, Whisking, heating by bain-marie, for storing a preparation or ingredients | |
150 mm | |
Flat bottom | |
11.4 L |
Stainless Steel | |
Fridge, deep-freezer, blast-chiller and oven | |
Each | |
Sterile |
Product Content Correction
Product Title
De Buyer™ Stainless Steel Pastry Bowls > Capacity: 11.4L