Learn More
Schneider Gerd™ Stainless Steel Pharmacists Spatula
Use this spatula for a variety of applications. Schneider Gerd™ Stainless Steel Pharmacists Spatulas are ideal for use with a wide range of compounds.
19.00€ - 20.00€
Specifications
Quantity | 1 Ea. |
---|---|
Product Type | Pharmacists Spatula |
Blade Material | Stainless Steel |
Handle Material | Wood |
Material | Stainless Steel |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10473464
|
Schneider Gerd™
611 |
210 mm |
19.00€
Each |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
10255004
|
Schneider Gerd™
613 |
260 mm |
20.00€
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
Description
- Spatulas are ideal for working with powders, stirring and mixing
- Perfect for miniscule amounts of chemicals
- Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
1 Ea. | |
Stainless Steel | |
Stainless Steel |
Pharmacists Spatula | |
Wood | |
Wood |