Learn More
Kap-Uts Culture Tube Closures
These Culture Tube Closures ensure the viability of microbiological cultures by protecting solid and broth media from excessive evaporation. They fit snugly on rimless culture tubes and flasks while allowing for gas exchange and ventilation.
646.00€ - 735.00€
Specifications
Product Type | Culture Tube Closure |
---|---|
Autoclavable | Autoclavable |
Material | Stainless Steel |
Quantity | 144 Pk. |
Tube Material | Stainless Steel |
Product Code | Brand | Size (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) | Price | Quantity & Availability | ||||||
15245376
|
Kap-Uts
04500-00 |
13 mm |
735.00€
Pack of 144 |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
15215637
|
Kap-Uts
04500-01 |
16 mm |
646.00€
Pack of 144 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
Description
- Closures minimize evaporation while allowing necessary gas exchange.
- Positive-grip closures of stainless steel or autoclavable polypropylene.
Specifications
Culture Tube Closure | |
Stainless Steel | |
Stainless Steel |
Autoclavable | |
144 Pk. |
Product Title
Kap-Uts Culture Tube Closures