Learn More
Sampling Systems™ Stainless Steel Sample Splitter Riffle Boxes
These easy-to-use Sample Splitters in stainless steel are for dividing and reducing bulk samples.
Brand:  Sampling Systems™ 8601S-06
 View more versions of this product
Product Code. 15879531
  • 626.00€ / Each
Only null left
Estimated Shipment: 23-07-2025
Description
  • Simply drop the sample into the top and the sample is split in two
  • Split sample drops into the riffle boxes underneath
  • It is recommended that width of slots are 2.5 to 3 times bigger than the diameter of the largest particle being split
  • Available in a wide range of sizes
Specifications
Specifications
Riffle Box
16
6 mm
Stainless Steel
Dividing and reducing bulk samples
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ Stainless Steel Sample Splitter Riffle Boxes > Slots: 16; Slot width: 6mm