Learn More
Swann-Morton™ Stainless Steel Scalpel Blades, 100-pack
Operate using Swann Morton™ Stainless Steel Scalpel Blades to ensure precision;
20.45€ - 59.50€
Specifications
Blade Material | Stainless Steel |
---|---|
Product Type | Scalpel Blade |
Certifications/Compliance | BS EN 7740 |
Product Code | Brand | For Use With (Equipment) | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Length (Metric) | Price | Quantity & Availability | |||||
12398009
|
Swann-Morton™
0301 |
The # 10 fits Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L | 25 mm |
59.50€
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
12981635
|
Swann-Morton™
0303 |
The # 11 Blade fit Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L | 20 mm |
53.60€
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
12991635
|
Swann-Morton™
0305 |
The # 15 Blade fit Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L | 15 mm |
20.45€
Pack of 100 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
12971635
|
Swann-Morton™
0302 |
Fitment # 3. Fits Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L | 15 mm |
20.45€
Pack of 100 |
Estimated Shipment: 04-04-2025
Log in to see stock. |
|||||
Description
- Sterile
- Stainless Steel
Specifications
Stainless Steel | |
Scalpel Blade | |
BS EN 7740 |
Product Title
Swann-Morton™ Stainless Steel Scalpel Blades, 100-pack