Learn More
Amsta Rostfritt Stainless-Steel Scoop
Brand:  Amsta Rostfritt 517550,5
Product Code. 11721776
  • 337.00€ / Each
Estimated Shipment: 07-08-2025
Specifications
Silver
316 mm
500 mL
Product Title
Amsta Rostfritt Stainless-Steel Scoop > Capacity: 500mL; Length: 316mm