Learn More
Nickel Electro™ Stainless Steel Gable Lid
Maintain stability in bath temperature and reduce evaporation with a stainless steel lid. Nickel Electro™ Clifton™ Stainless Steel Gable Lid minimises heat loss and reduces evaporation.
Brand:  Nickel Electro™ SL4-14
 View more versions of this product
Product Code. 11469519
  • 230.00€ / Each
Estimated Shipment: 04-07-2025
Description
  • Stainless steel
  • Insulated handle
  • Minimises heat loss
  • Reduces evaporation
  • Assists in maintaining bath temperature
Specifications
Specifications
Stainless steel gable lid
14 L Stirred Bath
Stainless Steel
3 Years
Documents
One moment while we fetch your results.
Product Title
Nickel Electro™ Stainless Steel Gable Lid > For Use With: 14L Bath