Learn More
Aalborg™ Precision Valve for Stainless Steel Flowmeter
Aalborg™ Precision Valve for Stainless Steel Flowmeter is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
214.00€ - 249.00€
Specifications
For Use With (Equipment) | 150 mm Gas/Liquid Flowmeter |
---|---|
Type | Standard Valve |
Product Code | Brand | Airflow Rate | Waterflow Rate | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Airflow Rate | Waterflow Rate | Price | Quantity & Availability | ||||
11778368
|
Aalborg™
VM3-SV |
1000 mL/min. | 26mL/min. |
214.00€
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
||||
11758368
|
Aalborg™
VM1-SV |
200 mL/min. | 6mL/min. |
249.00€
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
11768368
|
Aalborg™
VM2-SV |
400 mL/min. | 12mL/min. |
214.00€
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
11788368
|
Aalborg™
VM4-SV |
2500 mL/min. | 80mL/min. |
214.00€
Each |
Estimated Shipment: 11-03-2025
Log in to see stock. |
||||
11708378
|
Aalborg™
VM6-SV |
21500 mL/min. | 650mL/min. |
214.00€
Each |
Estimated Shipment: 11-03-2025
Log in to see stock. |
||||
Description
- For high precision
- Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
- They may also be used as shut off valves.
- Bodies of the valves are reinforced by structurally rigid metallic shells.
- Shells are made of stainless steel, bushings are made of plated brass or 316 stainless steel.
Specifications
150 mm Gas/Liquid Flowmeter | |
Standard Valve |