Learn More
Socorex™ Stainless Steel Straight Drenchers
Use Socorex™ Stainless Steel Straight Drenchers for animal feeding and dosing. These precision-assembled stainless-steel feeding needles are ideal for the delivery of fluid and are reusable since they are autoclavable.
33.90€ - 57.50€
Specifications
Product Type | Drenching Cannula |
---|---|
Autoclavable | Autoclavable |
Material | Stainless Steel |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11888432
|
Socorex™
376.0925S |
25 mm |
40.70€
Pack of 3 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
11838442
|
Socorex™
376.1251S |
51 mm |
50.55€
Pack of 3 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
11878442
|
Socorex™
376.16102S |
102 mm |
33.90€
Pack of 2 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
11873193
|
Socorex™
376.30203S |
203 mm |
57.50€
Pack of 2 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
Description
- Stainless Steel
- Various lengths to accommodate feeding needs
- Autoclavable
- Reusable
Specifications
Drenching Cannula | |
Autoclavable | |
Stainless Steel |