Learn More
Swann-Morton™ Stainless Steel Surgical Scalpels
Use for convenience in user environments: on the ward, in emergency or trauma units or even out in the field with paramedics. Swann Morton™ Stainless Steel Surgical Scalpels come in single peel packs.
Brand: Swann-Morton™ 0509
This item is not returnable.
View return policy
Description
- Stainless steel blade
- Polystyrene handle
- Individually wrapped
- Sterile
- Disposable
Specifications
Stainless Steel | |
Single-use | |
Polystyrene |
# 22A | |
Sterile |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 15
Product Content Correction
Product Title
Swann-Morton™ Stainless Steel Surgical Scalpels > No. 22A