Learn More
VACUUBRAND™ Stainless Steel T-Pieces With Small KF Flanges
124.00€ - 263.00€
Specifications
For Use With | Solenoid-operated valves VV and VV C |
---|---|
Material | Stainless Steel |
Quantity | 1 Each |
Type | T-piece |
Product Code | Brand | Diameter (Metric) Inside | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Price | Quantity & Availability | ||||||
10450463
|
VACUUBRAND™
20673514 |
16/16/16 mm |
145.00€
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
10059570
|
VACUUBRAND™
20673500 |
10/10/10 mm |
124.00€
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
10009600
|
VACUUBRAND™
20673515 |
25/25/25 mm |
184.00€
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
||||||
10682222
|
VACUUBRAND™
20673516 |
40/40/40 mm |
263.00€
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
Description
Specifications
Solenoid-operated valves VV and VV C | |
Stainless Steel | |
1 Each | |
T-piece |
Product Title
VACUUBRAND™ Stainless Steel T-Pieces With Small KF Flanges