Learn More
Linker Industrietechnik™ Stainless Steel Test Sieve, 200 mm OD, Nylon Sieve Skin
Constructed of durable stainless steel with a nylon sieve skin.
Brand:  Linker Industrietechnik™ 552.050.881-0028
 View more versions of this product
Product Code. 11765608
  • 247.00€ / Each
Estimated Shipment: 30-01-2025
Description
  • Stainless steel ring construction
  • Nylon sieve cloth/skin
  • Available in a variety of sieve mesh sizes
Specifications
Specifications
200 mm
Nylon
28 μm
Stainless Steel
28 μm