Learn More
Bochem™ Stainless Steel Tool Forceps
Enabling precision movement via self-tensing properties, these forceps have a sharp tip and a bent style.
Brand: Bochem™ 1700
This item is not returnable.
View return policy
Description
Self-tensing, sharp, bent forceps. Bochem™ Stainless Steel Tool Forceps are constructed of stainless steel with a plastic handle.
- Sharp tip
- Bent
- Plastic handle
- Self-tensing
Specifications
Sharp, Bent | |
160 mm | |
10/Pack | |
Self-tensing |
Stainless Steel | |
Forceps | |
Tool Forceps |
Product Content Correction
Product Title
Bochem™ Stainless Steel Tool Forceps > Tip Style: Sharp/Bent Tip; Length: 160mm