Learn More
Bochem™ Stainless Steel Tubing Clips
Used to control the flow of liquids. Bochem™ Stainless Steel Tubing Clips are manufactured from stainless steel.
Brand: Bochem™ 12132
This item is not returnable.
View return policy
Description
- Manufactured from stainless steel
- Used with tubing
Specifications
14 mm | |
Stainless Steel | |
Tubing Clip |
Product Content Correction
Product Title
Bochem™ Stainless Steel Tubing Clips > Diameter: 14mm