Learn More
Bochem™ Stainless Steel Tubing Clips
Used to control the flow of liquids. Bochem™ Stainless Steel Tubing Clips are manufactured from stainless steel.
Brand:  Bochem™ 12137
 View more versions of this product
Product Code. 10664032
  • 4.15€ / Each
Estimated Shipment: 07-05-2025
Description
  • Manufactured from stainless steel
  • Used with tubing
Specifications
Specifications
33 mm
Stainless Steel
Tubing Clip
Documents
One moment while we fetch your results.
Product Title
Bochem™ Stainless Steel Tubing Clips > Diameter: 33mm