Learn More
Koettermann™ Standard Hanging Cabinet, 1500x630x350mm
Koettermann™ Standard Hanging Cabinet, 1500x630x350mm provides suitable storage to accommodate a variety of laboratory needs.
544.00€ - 1035.00€
Specifications
Dimensions (D x W x H) | 350 x 1500 x 630 mm |
---|---|
No. of Shelves | 1 |
Product Type | Wall-mounted Cabinet |
Product Code | Brand | Locking Mechanism | Door Type | No. of Doors | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Locking Mechanism | Door Type | No. of Doors | Price | Quantity & Availability | |||
15733092
|
Koettermann™
307-00177 |
Yes | Sliding Glass Doors | 2 |
1035.00€
Each |
N/A | |||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
15743092
|
Koettermann™
307-00178 |
No | Open Front/No Doors | None |
544.00€
Each |
N/A | ||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
- Wall-mounted cabinet
- One shelf
- Side panels
Specifications
350 x 1500 x 630 mm | |
1 | |
Wall-mounted Cabinet |