Learn More
3M™ Standard Pipes
Brand:  3M™ 7000042677
256.01 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11701203
  • 463.00€ / Each
Estimated Shipment: 31-07-2025
Specifications
Standard Tube
1 Ea.
30 m
Documents
One moment while we fetch your results.
Product Title
3M™ Standard Pipes > Length: 30m