Learn More
3M™ Standard Pipes
Brand:  3M™ 7000042676
110.71 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11771143
  • 196.00€ / Each
Estimated Shipment: 31-07-2025
Specifications
Standard Pipe
1 Ea.
10 m
Product Title
3M™ Standard Pipes > Length: 10m