Learn More
Fisherbrand™ Standard Rotor Cover
GREENER_CHOICE
Provides protection from spinning contents in centrifuge. Fisherbrand™ Standard Rotor Cover should be placed on Mini Centrifuge prior to use.
Brand:  Fisherbrand™ HS100506
31.21 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Product Code. 12932051
  • 38.20€ / Each
Description
  • For use with Fisherbrand™ Mini-Centrifuge
  • Holds: 12 mL x 1.5 mL/2.0 mL Tube
  • Limits noise and aspiration that may escape from tubes
Specifications
Specifications
Plastic
Rotor Cover
Mini-Centrifuge
Product Title
Fisherbrand™ Standard Rotor Cover > For use with: Mini-Centrifuge