Learn More
Thermo Scientific Chemicals Standard Sample Tube, 3.24mm ID
For use with MKII Magnetic Susceptibility Balances
Brand: Thermo Scientific Chemicals 014349.KG
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- I.D.: 3.24mm
Specifications
1 Each | |
Standard Sample Tube |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A29U059 | Certificate of Analysis | 08/12/2021 | 014312.06,014312.14,014349.KG |
G06I18 | Certificate of Analysis | 08/12/2021 | 014312.06,014312.14,014349.KG |
D27K34 | Certificate of Analysis | 08/12/2021 | 014312.06,014312.14,014349.KG |
C01Q28 | Certificate of Analysis | 08/12/2021 | 014312.06,014312.14,014349.KG |
W10E019 | Certificate of Analysis | 08/12/2021 | 014312.06,014312.14,014349.KG |