Learn More
Standard Solution Bromide, WTW™
Product Code. 10325231
Click to view available options
Quantity:
1 L
Standard solutions to calibrate contains 10 g of the ion to be measured per 10 liters. By defined dilutions of these solutions, the user can easily produce his own calibration solution for the desired measuring range. The bulk with a one liter content makes for economic measurements.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 10g/L
Quantity 1 L
Boiling Point 100°C
Physical Form Fluid