missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5b Antibody (STAT5B/2611), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79907PE
This item is not returnable.
View return policy
Description
STAT5b Monoclonal antibody specifically detects STAT5b in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
STAT5b | |
Monoclonal | |
PE | |
signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
Recombinant full-length human STAT5b protein (Uniprot: P51692) | |
0.1 mL | |
Phospho Specific, Signal Transduction | |
6777 | |
Store at 4°C in the dark. | |
IgG1 Îş |
Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
STAT5B/2611 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
STAT5b Antibody (STAT5B/2611), PE, Novus Biologicals™ > 0.1 mL, PE
Spot an opportunity for improvement?Share a Content Correction