Learn More
Stearic acid glycidylester-d5, Honeywell Fluka™ Product Code.: 15970232
Stearic acid glycidylester-d5, Honeywell Fluka™
Product Code. 15970232 Shop All Honeywell Fluka Products
10 mg
Click to view available options
Quantity:
10 mg
Product Code. 15970232
Brand: Honeywell Fluka R148410MG
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
CAS 1346598-19-3
Packaging Glass Bottle
Quantity 10 mg
For Use With (Application) Dyes and Feed Additives
Physical Form Solid
Grade Analytical Standard
Product Content Correction
Product Title
Stearic acid glycidylester-d5, Honeywell Fluka™ >