Learn More
Justrite™ Steel Oily Waste Cans
Brand:  Justrite™ 09501
 View more versions of this product
Product Code. 11943649
  • 193.00€ / Each
Estimated Shipment: 30-05-2025
Specifications
52 L
SDS
Documents Certificates
Product Title
Justrite™ Steel Oily Waste Cans > Capacity: 52L