Learn More
Steel shot, non-spherical, 1mm (0.039in) dia, Thermo Scientific Chemicals
59.60€ - 107.54€
Specifications
Chemical Name or Material | Steel shot |
---|---|
Name Note | 1mm (0.039 in.) dia. |
MDL Number | MFCD00678174 |
Solubility Information | It is insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11339758
|
Thermo Scientific Alfa Aesar
044236.A3 |
2 kg |
59.60€
2kg |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
11349758
|
Thermo Scientific Alfa Aesar
044236.A7 |
5 kg |
107.54€
5kg |
In Stock
Log in to see stock. |
|||||
Description
Steel shot is used in cleaning applications for removal of loose material on metal surfaces. This type of cleaning is common in automotive industry (motor blocks, cylinder heads. Steel shot is also used in surface preparation process for cleaning metal surfaces which are covered with mill scale, dirt, rust, or paint coatings and for physically modifying the metal surface such as creating roughness for better application of paint and coating. The steel shots are generally employed in shot blasting machines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Steel shot | |
1mm (0.039 in.) dia. | |
MFCD00678174 | |
It is insoluble in water. |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only