Learn More
STEMCELL Technologies STEMdiff™ Cardiomyocyte Freezing Medium
Medium for cryopreserving hPSC-derived cardiomyocytes
Brand:  STEMCELL Technologies 05030
Encompass_Preferred
Product Code. 16337305
  • 253.80€ / 50mL
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • STEMdiff™ Cardiomyocyte Freezing Medium can be used to cryopreserve cardiomyocytes derived from human pluripotent stem cells (hPSCs) and maintained in STEMdiff™ Cardiomyocyte Maintenance Medium (Catalog #05020)
  • STEMdiff™ Cardiomyocyte Dissociation Kit (Catalog #05025) is required for dissociating cardiomyocytes prior to freezing
  • After thawing using STEMdiff™ Cardiomyocyte Support Medium (Catalog #05027), the cardiomyocytes can be used in various downstream applications and analyses
  • This medium supports hPSC-derived cardiomyocytes generated using STEMdiff™ Cardiomyocyte Differentiation Kit (Catalog #05010)
Specifications
Specifications
Cardiomyocytes, PSC-Derived
Cryopreservation
50 mL