missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ Neural Induction Medium

Encompass

Defined, serum-free medium for neural induction of human ES and iPS cells

492.00€ - 814.50€

Specifications

Cell Type Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Pluripotent Stem Cells
For Use With (Application) Cell Culture, Differentiation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17148301
Encompass
View Documents
STEMCELL Technologies
05835
250 mL
492.00€
250mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17158301
Encompass
View Documents
STEMCELL Technologies
05839
2 x 250 mL
814.50€
Pack of 2
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • STEMdiff™ Neural Induction Medium is a defined, serum-free medium for the neural induction of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells
  • This medium enables highly efficient generation of neural progenitor cell using either embryoid body- or monolayer culture-based protocols
Specifications

Specifications

Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Pluripotent Stem Cells
Cell Culture, Differentiation
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
STEMCELL Technologies STEMdiff™ Neural Induction Medium

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.