Learn More
Zeiss™ Stemi 305 Stereomicroscope with Flexible Stands
Compact Greenough stereo microscope with 5:1 zoom for biological education, labs and production environments.
5055.00€
Specifications
For Use With (Application) Biological Education, Labs and Production Environments
View More Specs
Products 1
Product Code Brand Base Style Price Quantity & Availability  
Product Code Brand Base Style Price Quantity & Availability  
17362830
View Documents
Zeiss™
491903-0011-000
Swiveling Stand U
5055.00€
1 set
Estimated Shipment: 12-03-2025
Description
  • This compact Greenough stereo microscope allows you to observe your samples as they are: three-dimensional and crisp in contrast and no preparation required.
  • The Stemi 305 is an easy-to-use microscope, requires minimal setup, everything is integrated including long-living LED illumination for both reflected and transmitted light.
  • Spot Illuminator K LED - Single spot for brightfield and darkfield epi-illumination, Thread M24x0.5 for polarizer, Height adjustable, inclinable, zoomable.
Specifications
Specifications
Biological Education, Labs and Production Environments