Learn More
Zeiss™ Stemi 305 Trinocular Stereo Microscope
Compact Greenough stereo microscope with 5:1 zoom for biological education, labs and production environments.
2495.00€ - 4420.00€
Specifications
Design Type Trinocular
View More Specs
Products 3
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
17100686
View Documents
Zeiss™
491903-0006-000
Double Spot Illuminator K LED
2665.00€
Each
17322830
View Documents
Zeiss™
491903-0007-000
Ring Illuminator K LED
2495.00€
1 set
Only null left
Estimated Shipment: 24-07-2025
Add to basket
17342830
View Documents
Zeiss™
491903-0009-000
Ring Illuminator VisiLED
4420.00€
1 set
Only null left
Estimated Shipment: 24-07-2025
Add to basket
Description
Description
  • This compact Greenough stereo microscope allows you to observe your samples as they are: three-dimensional and crisp in contrast and no preparation required.
  • The Stemi 305 is an easy-to-use microscope, requires minimal setup, everything is integrated including long-living LED illumination for both reflected and transmitted light.
  • Spot Illuminator K LED - Single spot for brightfield and darkfield epi-illumination, Thread M24x0.5 for polarizer, Height adjustable, inclinable, zoomable.
Specifications
Specifications
Trinocular
Documents
Product Certifications
Product Content Correction
Product Title
Zeiss™ Stemi 305 Trinocular Stereo Microscope